Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate ≥97%

Lieferant: Merck
Achtung

754730-1G
SIAL754730-1GEA 206 EUR
SIAL754730-1G
Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate ≥97%
Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate

Tris(2,2′-bipyridine)ruthenium(II) hexafluorophosphate is a high efficiency triplet emitter for OLED/sensor research.

Formel: C₃₀H₂₄F₁₂N₆P₂Ru
Schmelzpunkt: >300 °C
MDL: MFCD11042502
CAS-Nummer: 60804-74-2

Bestellen

Spezifikation Testergebnisse

Colour Orange to very dark orange, red to very dark red, orange-red and red-orange
Form Powder or crystal or chunk
Phosphorus NMR spectra Conforms to structure
Proton NMR spectrum Conforms to structure
Fluorine NMR spectra Conforms to structure
Carbon (C) 40,5 – 43,5%
Nitrogen (N) 9,4 – 10,1%
Assay 97%
Absorption 291 nm 80000 and 456 nm 13400


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR