DL-Ornithin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: H-DL-Orn-OH·HCl, 2,5-Diaminopentanoic acid hydrochloride, DL-Ornithine monohydrochloride

75490-10G
SIAL75490-10GEA 95.3 EUR
SIAL75490-10G
DL-Ornithin Hydrochlorid, Sigma-Aldrich®
DL-Ornithin Hydrochlorid
Formel: C₅H₁₂N₂O₂·HCl
Molecular Weight: 168.62 g/mol
MDL: MFCD00065398
CAS-Nummer: 1069-31-4
EINECS: 213-956-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR