(2R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Dexpanthenol, (R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamide

76200-100ML
SIAL76200-100MLEA 217 EUR
SIAL76200-100ML
(2R)-2,4-Dihydroxy-N-(3-hydroxypropyl)-3,3-dimethylbutanamid, Sigma-Aldrich®
D-Panthenol
Formel: C₉H₁₉NO₄
Molecular Weight: 205.25 g/mol
MDL: MFCD00065006
CAS-Nummer: 81-13-0
EINECS: 201-327-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR