Paraffin, liquid_highly, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Wax Paraffin, Paraffin wax

MQ100

76233-1L
SIAL76233-1LEA 71 EUR
SIAL76233-1L
Paraffin, liquid_highly, Sigma-Aldrich®
Paraffin
Siedepunkt: 300…450 °C (1013 hPa)
Dichte: 0.86 g/cm³ (20 °C)
MDL: MFCD00131611
CAS-Nummer: 8012-95-1
EINECS: 232-384-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR