Graphene oxide 2 mg/ml dispersion in water, Sigma-Aldrich®
Lieferant: Merck
763705-25ML
763705-100ML
SIAL763705-25MLEA
289
EUR
SIAL763705-25ML
SIAL763705-100ML
Graphene oxide 2 mg/ml dispersion in water, Sigma-Aldrich®
Graphene oxide
- Chloride free (purified by dialysis)
- Monolayer sheet
- Mean sheet diameter: 22 μm, 90% below 50 μm by laser diffraction
Features and Benefits: Large surface area, high chemical stability, good charge carrier properties.
Preparation Note: Product may aggregate over time to form larger particles. Sonicate before use if single layer graphene oxide required.
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...