Ammoniumsulfat 2 mol/l aqueous solution, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Diammoniumsulfat, di-Ammoniumsulfat

76399-100ML-F 76399-10ML-F
SIAL76399-100ML-FEA 219 EUR
SIAL76399-100ML-F SIAL76399-10ML-F
Ammoniumsulfat 2 mol/l aqueous solution, Sigma-Aldrich®
Ammoniumsulfat
Formel: (NH₄)₂SO₄
Molecular Weight: 132.14 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00003391
CAS-Nummer: 7783-20-2
EINECS: 231-984-1
Merck Index: 13,00559

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR