Xenon 99,99, Messer® CANGas

Lieferant: Merck

769118-1L
SIAL769118-1LEA 1230 EUR
SIAL769118-1L
Xenon 99,99, Messer® CANGas
Xenon

Xenon is a colourless and odourless rare gas that is heavier than air.


It is used for filling gas for lamps (halogen lamps, gas discharge lamps) and anesthetic gas in medicine.


Xenon is also a component of excimer laser gas mixtures and panning gas for plasma display panels (PDP). Gas is easy to handle in a small, light pressure can, while the disposable container allows for aluminium recycling when finished.

Formel: Xe
Molecular Weight: 131.29 g/mol
Siedepunkt: -108.1
Schmelzpunkt: -111.9
Dichte: 0.005366
MDL: MFCD00036280
CAS-Nummer: 7440-63-3
UN: 2036
ADR: 2
Merck Index: 12,10206

Bestellen

Spezifikation Testergebnisse

CAS Number 7440-63-3
Empirical Formula (Hill Notation) Xe
Molecular Weight 131,29
EC Number 231-172-7
MDL number MFCD00036280
PubChem Substance ID 329767434
Filling mass 71 g
Filling volume 12 l
InChI Key FHNFHKCVQCLJFQ-UHFFFAOYSA-N
Assay 99,99%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR