3-Pentensäure, Sigma-Aldrich®

Lieferant: Merck

77047-5ML
SIAL77047-5MLEA 102 EUR
SIAL77047-5ML
3-Pentensäure, Sigma-Aldrich®
3-Pentenoic acid
Formel: C₅H₈O₂
Molecular Weight: 100.12 g/mol
CAS-Nummer: 5204-64-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR