2-Brom-1-ethylpyridiniumtetrafluoroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Bromo-1-ethyl-pyridinium tetrafluoroborate, 2-Bromo-1-ethylpyridin-1-ium tetrafluoroborate

77386-5G-F
SIAL77386-5G-FEA 136 EUR
SIAL77386-5G-F
2-Brom-1-ethylpyridiniumtetrafluoroborat, Sigma-Aldrich®
2-Bromo-1-ethylpyridinium tetrafluoroborate
Formel: C₇H₉BBrF₄N
Molecular Weight: 273,86 g/mol
MDL: MFCD00060046
CAS-Nummer: 878-23-9
EINECS: 212-900-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR