3-Phenoxytoluol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Phenyl-m-tolylether

77780-100ML
SIAL77780-100MLEA 145 EUR
SIAL77780-100ML
3-Phenoxytoluol, Sigma-Aldrich®
3-Phenoxytoluol
Formel: C₁₃H₁₂O
Molecular Weight: 184,24 g/mol
Siedepunkt: 271 °C (1013 hPa)
Dichte: 1,051 g/cm³ (25 °C)
MDL: MFCD00008531
CAS-Nummer: 3586-14-9
EINECS: 222-716-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR