(R)-(-)-1-Methoxy-2-propanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (R)-(-)-Propylenglycolmonomethylether

77913-5ML-F
SIAL77913-5ML-FEA 0 EUR
SIAL77913-5ML-F
(R)-(-)-1-Methoxy-2-propanol, Sigma-Aldrich®
(R)-(-)-1-Methoxy-2-propanol
Formel: C₄H₁₀O₂
Molecular Weight: 90,12 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01632587
CAS-Nummer: 4984-22-9
UN: 3092
ADR: 3,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR