Methyl-β-cyclodextrin (mixture of several Methylated), Sigma-Aldrich®
Lieferant: Merck
779873-100G
779873-10KG
779873-1KG
779873-500G
SIAL779873-100GEA
483
EUR
SIAL779873-100G
SIAL779873-10KG
SIAL779873-1KG
SIAL779873-500G
Methyl-β-cyclodextrin (mixture of several Methylated), Sigma-Aldrich®
Methyl-beta-cyclodextrin (mixture of several Methylated)
Methyl-β-cyclodextrin is a macrocyclic compound that can form inclusion complexes with a number of guest molecules. It shows higher solubility in aqueous solutions and greater solubilising and complexing power compared to the parent β-cyclodextrin.
MβCD can be used to enhance the enzyme activity and enantioselectivity of subtilisin enzyme in organic solvents.
Produced by Wacker Chemie AG.
Flammpunkt:
187 °C |
MDL:
MFCD00074980 CAS-Nummer: 128446-36-6 |
Spezifikation Testergebnisse
Assay | ≥98.0% Cyclodextrin Basis |
Appearance (Colour) | White to Off White |
Appearance (Form) | Powder or Crystals |
Halides (as Chloride) | ≤0.20% |
Specific Rotation (20/D) | 154.0 - 165.0 Degrees |
Loss on Drying | ≤5.0% |
Residue on Ignition | ≤0.50% |
Infrared Spectrum | Conforms to Structure |
UV Method | 10% in H₂O |
Reducing Substances | ≤0.50% |
Volatile Impurities | Volatile organics ≤50 ppm |
Absorbance, 220 - 350 nm | ≤1.0 |
Absorbance, 350 - 650 nm | ≤0.10 |
Degree of Substitution | Molar Substitution 1.7 - 1.9 |
Heavy Metals | ≤10 ppm |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...