N,N-Bis(trifluormethansulfonyl)anilin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N-Phenylbis(trifluoromethanesulphonimide)

78175-25G 78175-50G 78175-5G
SIAL78175-25GEA 176 EUR
SIAL78175-25G SIAL78175-50G SIAL78175-5G
N,N-Bis(trifluormethansulfonyl)anilin, Sigma-Aldrich®
N,N-Bis(trifluormethansulfonyl)anilin
Formel: C₈H₅F₆NO₄S₂
MDL: MFCD00000404
CAS-Nummer: 37595-74-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR