1-Butyl-2,3-dimethylimidazoliumchlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Butyl-2,3-dimethyl-3-imidazolium chloride

78194-50G-F
SIAL78194-50G-FEA 166 EUR
SIAL78194-50G-F
1-Butyl-2,3-dimethylimidazoliumchlorid, Sigma-Aldrich®
1-Butyl-2,3-dimethylimidazolium chloride
Formel: C₉H₁₇ClN₂
Molecular Weight: 188,7 g/mol
MDL: MFCD03093291
CAS-Nummer: 98892-75-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR