p-Phenylendiamin Dihydrochlorid, Sigma-Aldrich®
Lieferant: Merck
|
Gefahr
|
78460-100G
SIAL78460-100GEA
123
EUR
SIAL78460-100G
p-Phenylendiamin Dihydrochlorid, Sigma-Aldrich®
p-Phenylendiamin Dihydrochlorid
Formel:
C₆H₁₀Cl₂N₂ Schmelzpunkt: 275 °C Dichte: >1 g/cm³ (20 °C) |
MDL:
MFCD00013002 CAS-Nummer: 624-18-0 EINECS: 210-834-9 UN: 2811 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...