Phenylpropargylether, Sigma-Aldrich®

Lieferant: Merck

Synonyme: (2-Propinyloxy)benzol

78960-10ML
SIAL78960-10MLEA 101 EUR
SIAL78960-10ML
Phenylpropargylether, Sigma-Aldrich®
Phenylpropargylether
Formel: C₉H₈O
Molecular Weight: 132,16 g/mol
MDL: MFCD00048107
CAS-Nummer: 13610-02-1
EINECS: 237-095-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR