(±)-1-Phenyl-2,2,2-trifluorethanol, Sigma-Aldrich®

Lieferant: Merck

Synonyme: α-(Trifluormethyl)benzylalkohol

79234-5ML
SIAL79234-5MLEA 72.8 EUR
SIAL79234-5ML
(±)-1-Phenyl-2,2,2-trifluorethanol, Sigma-Aldrich®
(±)-1-Phenyl-2,2,2-trifluorethanol
Formel: C₆H₅CH(OH)CF₃
Molecular Weight: 176,14 g/mol
MDL: MFCD00004498
CAS-Nummer: 340-05-6
EINECS: 206-429-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR