Methylamine hydroiodide ≥98%, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Methylammonium iodide

793493-5G
SIAL793493-5GEA 252 EUR
SIAL793493-5G
Methylamine hydroiodide ≥98%, Sigma-Aldrich®
Methylamine hydroiodide

Methanamine hydriodide is an important precursor for the preparation of perovskite photoactive layers for solar energy conversion.

Formel: CH₃NH₂·HI
Schmelzpunkt: 270,..280 °C
MDL: MFCD28100833
CAS-Nummer: 14965-49-2

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Yellow
Appearance (Form) Powder or Crystals
Proton NMR Spectrum Conforms to Structure
Titration with AgNO₃ 97.5 - 102.5%
Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR