2,5-Dibrompyrazin 95% (durch GC)

Lieferant: Merck
Achtung

796603-1G
SIAL796603-1GEA 37.9 EUR
SIAL796603-1G
2,5-Dibrompyrazin 95% (durch GC)
2,5-Dibromopyrazine
Formel: C₄H₂Br₂N₂
Molecular Weight: 237,8800 g/mol
MDL: MFCD08061601
CAS-Nummer: 23229-26-7

Bestellen

Spezifikation Testergebnisse

Appearance (Colour) White to Yellow and Faint Beige to Beige and Faint Brown to Brown
Appearance (Form) Powder or Crystals
Infrared Spectrum Conforms to Structure
Purity (GC) ≥94.5%
Carbon 18.1 - 22.3%
Nitrogen 10.5 - 13.0%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR