Sulpho-EGS (Ethylene glycol bis(sulphosuccinimidyl succinate)), Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Sulfo EGS

M-Clarity™ Qualitätsstufe = MQ100

803219-50MG
SIAL803219-50MGEA 250 EUR
SIAL803219-50MG
Sulpho-EGS (Ethylene glycol bis(sulphosuccinimidyl succinate)), Sigma-Aldrich®
Sulpho-EGS (Ethylene glycol bis(sulphosuccinimidyl succinate))

EGS is a water-insoluble, homobifunctional N-hydroxysuccinimide ester (NHS ester) and Sulfo-EGS is its water-soluble analog. The spacer arm contains two cleavable ester sites that may be broken with hydroxylamine, which yields two fragments with terminal amide bonds and the release of ethylene glycol. These reagents are often used for conjugating radiolabeled ligands to cell surface receptors. The water insoluble reagent EGS does not possess a charged group, is lipophilic and, therefore, membrane-permeable and useful for intracellular and intramembrane protein conjugation. EGS is first dissolved in DMSO or DMF and added to the aqueous reaction mixture at a final solvent concentration of 10-20%, to minimize detrimental affects to the protein.


  • Reactive groups: sulfo-NHS ester (both ends)
  • Reactive towards: amino groups (primary amines)
  • Water-soluble; compare to EGS
  • Membrane-impermeable, allowing for cell surface labeling
  • Crosslinks formed are reversible at pH 8.5 using hydroxylamine for 3 to 6 hours at 37°C
  • Lactose dehydrogenase retained 60% of its activity after reversible crosslinking with EGS


Caution
This product is sensitive to moisture. The vial is packaged in a resealable bag with a desiccant to reduce exposure to moisture. After cold storage, equilibrate the vial to room temperature before opening to reduce condensation inside the vial. Make fresh solutions. Storage of stock solutions is not recommended. After use, return the vial to the resealable bag. Close the bag and store the product at the recommended temperature.

Formel: C₁₈H₁₈O₁₈N₂Na₂S₂
Molecular Weight: 660,45 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00054988
CAS-Nummer: 67410-92-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR