Sulpho-SANPAH (Sulphosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate), Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: Sulfo SANPAH

M-Clarity™ Qualitätsstufe = MQ100

803332-50MG
SIAL803332-50MGEA 401 EUR
SIAL803332-50MG
Sulpho-SANPAH (Sulphosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate), Sigma-Aldrich®
Sulpho-SANPAH (Sulphosuccinimidyl-6-(4′-azido-2′-nitrophenylamino)hexanoate)

ANB-NOS and Sulfo-SANPAH are heterobifunctional crosslinkers that contain an amine-reactive N-hydroxysuccinimide (NHS) ester and a photoactivatable nitrophenyl azide. NHS esters react efficiently with primary amino groups (-NH2) in pH 7-9 buffers to form stable amide bonds. When exposed to UV light nitrophenyl azides form a nitrene group that can initiate addition reactions with double bonds, insertion into C-H and N-H sites, or subsequent ring expansion to react with a nucleophile (e.g., primary amines).


  • Reactive groups: sulfo-NHS ester and nitrophenyl azide
  • Reactive towards: amino groups and any nucleophile
  • Non-cleavable
  • N-Sulfosuccinimidyl-6-(4′-azido-2′-nitrophenylamino) hexanoate
  • Optimal photolysis occurs at 320-350 nm; minimizes damage to biomolecules by irradiation
  • Water soluble; non-cleavable


Bitte beachten This product is sensitive to moisture. The vial is packaged in a resealable bag with a desiccant to reduce exposure to moisture. After cold storage, equilibrate the vial to room temperature before opening to reduce condensation inside the vial. Make fresh solutions. Storage of stock solutions is not recommended. After use, return the vial to the resealable bag. Close the bag and store the product at the recommended temperature.

Formel: C₁₆H₁₇O₉N₆NaS
Molecular Weight: 492,40 g/mol
Lagertemperatur: Gefrierschrank
CAS-Nummer: 102568-43-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR