BMPH (N-(β-maleimidopropionic acid) hydrazide trifluoroacetic acid salt) ≥90%, Sigma-Aldrich®

Lieferant: Merck

Synonyme: BMPH

M-Clarity™ Qualitätsstufe = MQ100

803529-50MG
SIAL803529-50MGEA 206 EUR
SIAL803529-50MG
BMPH (N-(β-maleimidopropionic acid) hydrazide trifluoroacetic acid salt) ≥90%, Sigma-Aldrich®
BMPH (N-(β-maleimidopropionic acid) hydrazide trifluoroacetic acid salt)

BMPH, EMCH and KMUH are heterobifunctional crosslinkers containing sulfhydryl-reactive maleimide and carbonyl-reactive hydrazide moieties. Maleimides react with free sulfhydryls (-SH) to form stable thioether bonds. Hydrazide groups react with carbonyls (aldehydes and ketones) to form stable hydrazone bonds. Aldehyde groups can be created by periodate-oxidation of sialic acid and other sugar components of glycoprotein polysaccharides. Thus, these crosslinkers are useful for conjugating glycoproteins and sulfhydryl-containing peptides or proteins. Alternatively, the amine of the hydrazide moiety can be reacted to carboxyl groups using the crosslinker EDC.


  • Reactive groups: maleimide and hydrazide
  • Reactive towards: sulfhydryl groups and carbonyl (aldehyde) groups
  • Short (8.1A), sulfhydryl-to-aldehyde crosslinker with simple spacer arm (noncleavable)
  • Maleimide group reacts with sulfhydryl groups to form stable thioether linkages
  • Hydrazide group conjugates to oxidized sugars of glycoproteins and carbohydrates
  • Use sodium meta-periodate to oxidize glycosylation (e.g., sialic acid) to reactive aldehyde groups
  • Use with EDC to conjugate primary amine of hydrazide group to carboxyl groups


Bitte beachten This product is sensitive to moisture. The vial is packaged in a resealable bag with a desiccant to reduce exposure to moisture. After cold storage, equilibrate the vial to room temperature before opening to reduce condensation inside the vial. Make fresh solutions. Storage of stock solutions is not recommended. After use, return the vial to the resealable bag. Close the bag and store the product at the recommended temperature.

Formel: C₇H₉N₃O₃·CF₃CO₂H
Molecular Weight: 297,19 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01863410
CAS-Nummer: 359436-61-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR