DTME (dithio-bis-maleimidoethane) ≥90%, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: DTME

M-Clarity™ Qualitätsstufe = MQ100

803618-50MG
SIAL803618-50MGEA 446 EUR
SIAL803618-50MG
DTME (dithio-bis-maleimidoethane) ≥90%, Sigma-Aldrich®
DTME (dithio-bis-maleimidoethane)

DTME is a homobifunctional, maleimide crosslinker for conjugation between sulfhydryl groups (-SH). Such bismaleimide crosslinkers are commonly used to explore and characterize protein structure (i.e., oligomerization) or protein interactions. DTME is similar in length to BMH but differs in containing a disulfide bond in its spacer arm, allowing crosslinks with DTME to be cleaved with reducing agent such as dithiothreitol (DTT).


  • Reactive groups: maleimide (both ends)
  • Reactive towards: sulfhydryl groups
  • Long (13.3A), cleavable, sulfhydryl-to-sulfhydryl crosslinker, composed of maleimide groups and 11-atom disulfide spacer arm
  • Water-insoluble—dissolve first in DMF or DMSO, then add to aqueous reaction buffers
  • Cleavable by reduction of disulfide spacer arm with DTT, TCEP or other reducing agent


Bitte beachten This product is sensitive to moisture. The vial is packaged in a resealable bag with a desiccant to reduce exposure to moisture. After cold storage, equilibrate the vial to room temperature before opening to reduce condensation inside the vial. Make fresh solutions. Storage of stock solutions is not recommended. After use, return the vial to the resealable bag. Close the bag and store the product at the recommended temperature.

Formel: C₁₂H₁₂N₂O₄S₂
Molecular Weight: 312,36 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01863385
CAS-Nummer: 71865-37-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR