SMPH (Succinimidyl-6-((beta-maleimidopropionamido)hexanoate) ≥90%, Sigma-Aldrich®

Lieferant: Merck
Achtung

Synonyme: SMPH

M-Clarity™ Qualitätsstufe = MQ100

803626-50MG
SIAL803626-50MGEA 314 EUR
SIAL803626-50MG
SMPH (Succinimidyl-6-((beta-maleimidopropionamido)hexanoate) ≥90%, Sigma-Aldrich®
SMPH (Succinimidyl-6-((beta-maleimidopropionamido)hexanoate)

SMPH is a heterobifunctional crosslinker with N-hydroxysuccinimide (NHS) ester and maleimide groups that allow covalent conjugation of amine- and sulfhydryl-containing molecules. NHS esters react with primary amines at pH 7-9 to form amide bonds, while the maleimides react with sulfhydryl groups at pH 6.5-7.5 to form stable thioether bonds. In aqueous solutions, hydrolytic degradation of the NHS ester is a competing reaction whose rate increases with pH. The maleimide group is more stable than the NHS-ester group but will slowly hydrolyze and also lose its reaction specificity for sulfhydryls at pH values greater than 7.5. For these reasons, conjugation experiments involving this type of heterobifunctional crosslinker are usually performed at pH 7.2-7.5, with the NHS-ester (amine-targeted) reaction being accomplished before or simultaneous with the maleimide (sulfhydryl-targeted) reaction.


  • Reactive groups: NHS ester and maleimide
  • Reactive towards: amino and sulfhydryl groups
  • Slightly longer and more soluble than SMPB
  • NHS ester reacts with primary amines at pH 7-9, forming a stable amide bond
  • Maleimide reacts with -SH groups at pH 6.5-7.5, forming a stable thioether bond
  • Non-cleavable
  • Water-insoluble (dissolve first in DMF or DMSO)


Bitte beachten This product is sensitive to moisture. The vial is packaged in a resealable bag with a desiccant to reduce exposure to moisture. After cold storage, equilibrate the vial to room temperature before opening to reduce condensation inside the vial. Make fresh solutions. Storage of stock solutions is not recommended. After use, return the vial to the resealable bag. Close the bag and store the product at the recommended temperature.

Formel: C₁₇H₂₁N₃O₇
Molecular Weight: 379.36 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD01863460
CAS-Nummer: 367927-39-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR