Propargylbromid 80% ∼9.2 mol/l in Toluol ≥80%, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
3-Brompropin
M-Clarity™ Qualitätsstufe = MQ200
81831-250ML
81831-50ML
SIAL81831-250MLEA
385
EUR
SIAL81831-250ML
SIAL81831-50ML
Propargylbromid 80% ∼9.2 mol/l in Toluol ≥80%, Sigma-Aldrich®
Propargylbromid
Formel:
C₃H₃Br Molecular Weight: 118.96 g/mol Siedepunkt: 90 Schmelzpunkt: -43 |
MDL:
MFCD00000241 CAS-Nummer: 106-96-7 EINECS: 203-447-1 UN: 2924 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...