4-(1-Pyrenyl)buttersäurehydrazid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Pyren-1-yl)butanehydrazide

M-Clarity™ Qualitätsstufe = MQ100

82669-100MG
SIAL82669-100MGEA 237 EUR
SIAL82669-100MG
4-(1-Pyrenyl)buttersäurehydrazid, Sigma-Aldrich®
4-(1-Pyrenyl)buttersäurehydrazid
Formel: C₂₀H₁₈N₂O
Molecular Weight: 302.3697 g/mol
MDL: MFCD00467260
CAS-Nummer: 55486-13-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR