Pyridinium-p-toluolsulfonat, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

82815-100G 82815-25G
SIAL82815-100GEA 137 EUR
SIAL82815-100G SIAL82815-25G
Pyridinium-p-toluolsulfonat, Sigma-Aldrich®
Pyridinium-p-toluolsulfonat
Formel: C₁₂H₁₃NO₃S
MDL: MFCD00013108
CAS-Nummer: 24057-28-1
EINECS: 246-002-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR