Magnesiumsulfat 2.5 mol/l aqueous solution, Sigma-Aldrich®

Lieferant: Merck

Synonyme: MgSO4, Magnesium sulphate anhydrous

MQ200

83266-100ML-F 83266-500ML-F
SIAL83266-100ML-FEA 87.9 EUR
SIAL83266-100ML-F SIAL83266-500ML-F
Magnesiumsulfat 2.5 mol/l aqueous solution, Sigma-Aldrich®
Magnesium sulphate
Formel: MgSO₄
Molecular Weight: 120.37 g/mol
Schmelzpunkt: 1127
Lagertemperatur: Raumtemperatur
MDL: MFCD00011110
CAS-Nummer: 7487-88-9
EINECS: 231-298-2
Merck Index: 12,05731
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bc2ae189b0cbc1bfc599951528466503d0d9008&vid=63867221020ebd2c8c227492e609a4764639ff79&prid=web&clid=SAPDAM

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR