Salicylaldoxim, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 2-Hydroxybenzaldehyde oxime

84172-100G
SIAL84172-100GEA 166 EUR
SIAL84172-100G
Salicylaldoxim, Sigma-Aldrich®
Salicylaldoxim
Formel: C₇H₇NO₂
MDL: MFCD00002120
CAS-Nummer: 94-67-7
EINECS: 202-353-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR