Schwefeltrioxid-Pyridin-Komplex, Sigma-Aldrich®
Lieferant: Merck
84737-100G
84737-500G
SIAL84737-100GEA
86.4
EUR
SIAL84737-100G
SIAL84737-500G
Schwefeltrioxid-Pyridin-Komplex, Sigma-Aldrich®
Schwefeltrioxid-Pyridin-Komplex
Formel:
C₅H₅NO₃S |
MDL:
MFCD00012437 CAS-Nummer: 26412-87-3 EINECS: 247-683-3 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...