5-Bromuridin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

850187-1G 850187-250MG
SIAL850187-1GEA 96.2 EUR
SIAL850187-1G SIAL850187-250MG
5-Bromuridin, Sigma-Aldrich®
5-Bromuridin
Formel: C₉H₁₁BrN₂O₆
Molecular Weight: 323.10 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00006528
CAS-Nummer: 957-75-5
EINECS: 213-486-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR