5-Bromuridin, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
850187-1G
850187-250MG
SIAL850187-1GEA
96.2
EUR
SIAL850187-1G
SIAL850187-250MG
5-Bromuridin, Sigma-Aldrich®
5-Bromuridin
Formel:
C₉H₁₁BrN₂O₆ Molecular Weight: 323.10 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00006528 CAS-Nummer: 957-75-5 EINECS: 213-486-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...