(3,4-Dihydroxyphenyl)essigsäure, Sigma-Aldrich®
Lieferant: Merck
|
Achtung
|
Synonyme:
DOPAC, 3,4-Dihydroxybenzeneacetic acid, 3,4-Dihydroxyphenylacetic acid
M-Clarity™ Qualitätsstufe = MQ100
850217-1G
850217-5G
SIAL850217-1GEA
47.1
EUR
SIAL850217-1G
SIAL850217-5G
(3,4-Dihydroxyphenyl)essigsäure, Sigma-Aldrich®
(3,4-Dihydroxyphenyl)essigsäure
Formel:
(C₆H₃(OH)₂)CH₂COOH Molecular Weight: 168.15 g/mol Siedepunkt: 337 Schmelzpunkt: 131…132 °C |
MDL:
MFCD00004338 CAS-Nummer: 102-32-9 EINECS: 203-024-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...