1,2-Distearoyl-sn-glycero-3-phosphocholine ≥99% (durch TLC)

Lieferant: Merck

Synonyme: DSPC, 1,2-Distearoyl-sn-glycero-3-phosphorylcholine, 1,2-Dioctadecanoyl-sn-glycero-3-phosphocholine, (R)-2,3-Bis(stearoyloxy)propyl [2-(Trimethylammonio)ethyl] Phosphate, (R)-2,3-Bis(stearoyloxy)propyl (2-(trimethylammonio)ethyl) phosphate

850365P-500MG
SIAL850365P-500MGEA 157 EUR
SIAL850365P-500MG
1,2-Distearoyl-sn-glycero-3-phosphocholine ≥99% (durch TLC)
1,2-Distearoyl-sn-glycero-3-phosphocholine
Formel: C₄₄H₈₈NO₈P
Molecular Weight: 790,16 g/mol
Lagertemperatur: Gefrierschrank
MDL: MFCD00036905
CAS-Nummer: 816-94-4

Bestellen

Spezifikation Testergebnisse

TLC, Purity >99%
TLC, Ninhydrin Negative
TLC, Iodine One Major Spot
TLC, Phosphorus Positive
TLC, Charring Negative
TLC, Water Dip One Major Spot
FAME (GC/FID), NLT 99% 18:0 (AUC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR