5-Azacytosin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-Amino-1,3,5-triazin-2(1H)-one, 6-Amino-1,3,5-triazin-2(1H)-one, 2-Amino-4-hydroxy-1,3,5-triazine

M-Clarity™ Qualitätsstufe = MQ200

855049-5G
SIAL855049-5GEA 71 EUR
SIAL855049-5G
5-Azacytosin, Sigma-Aldrich®
5-Azacytosin
Formel: C₃H₄N₄O
Molecular Weight: 112,09 g/mol
MDL: MFCD00149402
CAS-Nummer: 931-86-2
EINECS: 213-242-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR