5-Amino-2,4(1H,3H)-pyrimidindion, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Aminouracil, 5-Amino-2,4-dihydroxypyrimidin
M-Clarity™ Qualitätsstufe = MQ200
855286-25G
SIAL855286-25GEA
147
EUR
SIAL855286-25G
5-Amino-2,4(1H,3H)-pyrimidindion, Sigma-Aldrich®
5-Aminouracil
Formel:
C₄H₅N₃O₂ Molecular Weight: 127,1 g/mol Siedepunkt: 377 °C (1013 hPa) Schmelzpunkt: 300 °C |
MDL:
MFCD00006025 CAS-Nummer: 932-52-5 EINECS: 213-252-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...