Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin), Sigma-Aldrich®

Lieferant: Merck

Synonyme: N1,N1'-(Butane-1,4-diyl)bis(propane-1,3-diamine), Spermine

M-Clarity™ Qualitätsstufe = MQ200

85590-25G 85590-5G
SIAL85590-25GEA 757 EUR
SIAL85590-25G SIAL85590-5G
Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin), Sigma-Aldrich®
Spermin (N,N'-Bis(3-aminopropyl)-1,4-butandiamin)
Formel: C₁₀H₂₆N₄
MDL: MFCD00008215
CAS-Nummer: 71-44-3
EINECS: 200-754-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR