β-Cyclodextrin Hydrat, Sigma-Aldrich®

Lieferant: Merck


MQ200

856088-25G 856088-100G 856088-5G
SIAL856088-25GEA 58.1 EUR
SIAL856088-25G SIAL856088-100G SIAL856088-5G
β-Cyclodextrin Hydrat, Sigma-Aldrich®
β-Cyclodextrin Hydrat
Formel: C₄₂H₇₀O₃₅·H₂O
Molecular Weight: 1134.98 g/mol (anhy)
MDL: MFCD00150811
CAS-Nummer: 68168-23-0
EINECS: 231-493-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR