4-Brom-DL-phenylalanin, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

857246-1G
SIAL857246-1GEA 35.8 EUR
SIAL857246-1G
4-Brom-DL-phenylalanin, Sigma-Aldrich®
4-Brom-DL-phenylalanin
Formel: C₉H₁₀BrNO₂
Molecular Weight: 244,09 g/mol
MDL: MFCD00002599
CAS-Nummer: 14091-15-7
EINECS: 237-941-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR