m-Kresolpurpur, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ200
857890-10G
857890-1G
857890-5G
SIAL857890-10GEA
102
EUR
SIAL857890-10G
SIAL857890-1G
SIAL857890-5G
m-Kresolpurpur, Sigma-Aldrich®
m-Kresolpurpur
Formel:
C₂₁H₁₈O₅S |
MDL:
MFCD00005871 CAS-Nummer: 2303-01-7 EINECS: 218-960-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...