m-Kresolpurpur, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ200

857890-10G 857890-1G 857890-5G
SIAL857890-10GEA 102 EUR
SIAL857890-10G SIAL857890-1G SIAL857890-5G
m-Kresolpurpur, Sigma-Aldrich®
m-Kresolpurpur
Formel: C₂₁H₁₈O₅S
MDL: MFCD00005871
CAS-Nummer: 2303-01-7
EINECS: 218-960-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR