3,4-Dihydroxybenzylamin Hydrobromid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 4-(Aminomethyl)benzene-1,2-diol hydrobromide

M-Clarity™ Qualitätsstufe = MQ200

858781-250MG 858781-1G 858781-5G
SIAL858781-250MGEA 52.8 EUR
SIAL858781-250MG SIAL858781-1G SIAL858781-5G
3,4-Dihydroxybenzylamin Hydrobromid, Sigma-Aldrich®
3,4-Dihydroxybenzylamin Hydrobromid
Formel: C₇H₁₀BrNO₂
Molecular Weight: 220.07 g/mol
MDL: MFCD00012859
CAS-Nummer: 16290-26-9
EINECS: 240-382-8

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR