6-Fluortryptamin Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

860255-500MG
SIAL860255-500MGEA 332 EUR
SIAL860255-500MG
6-Fluortryptamin Hydrochlorid, Sigma-Aldrich®
6-Fluortryptamin Hydrochlorid
Formel: C₁₀H₁₂ClFN₂
Molecular Weight: 214,67 g/mol
MDL: MFCD00012687
CAS-Nummer: 55206-24-1
EINECS: 259-533-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR