L-Methioninmethylester Hydrochlorid, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
H-Met-OMe.HCl, H-Met-OMe·HCl
M-Clarity™ Qualitätsstufe = MQ200
860409-25G
SIAL860409-25GEA
208
EUR
SIAL860409-25G
L-Methioninmethylester Hydrochlorid, Sigma-Aldrich®
L-Methioninmethylester Hydrochlorid
Formel:
C₆H₁₃NO₂S·HCl Molecular Weight: 199,7 g/mol |
MDL:
MFCD00012491 CAS-Nummer: 2491-18-1 EINECS: 219-651-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...