3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Dihydrochloride TMB, TMB-HCl
M-Clarity™ Qualitätsstufe = MQ200
861510-1G
SIAL861510-1GEA
107
EUR
SIAL861510-1G
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat, Sigma-Aldrich®
3,3',5,5'-Tetramethylbenzidin Dihydrochlorid Hydrat
Formel:
C₁₆H₂₀N₂·2HCl·xH₂O Molecular Weight: 313,27 g/mol (anhy) |
MDL:
MFCD00150104 CAS-Nummer: 207738-08-7 EINECS: 264-769-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...