Sodium taurocholate hydrate ≥97,0% (durch TLC), Sigma-Aldrich®

Lieferant: Merck

Synonyme: Taurocholic acid sodium salt hydrate, Cholaic acid sodium salt hydrate, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate hydrate

86339-1G 86339-25G 86339-5G
SIAL86339-1GEA 63.4 EUR
SIAL86339-1G SIAL86339-25G SIAL86339-5G
Sodium taurocholate hydrate ≥97,0% (durch TLC), Sigma-Aldrich®
Sodium taurocholate hydrate

Sodium taurocholate hydrate is used as an emulsifier in in situ preparation of digestive enzymes.

Formel: C₂₆H₄₆NNaO₈S
Molecular Weight: 555.70 g/mol
MDL: MFCD00150819
CAS-Nummer: 345909-26-4

Bestellen

Spezifikation Testergebnisse

Assay (TLC) ≥97,0%
Optical activity [α]20/D +23±1°, c= 3% in H₂O (dry matter)
Impurities (taurine) ≤1%
Impurities (cholic acid) ≤2%
Solubility in H₂O 50 mg/ml clear


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR