Sodium taurocholate hydrate ≥97,0% (durch TLC), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Taurocholic acid sodium salt hydrate, Cholaic acid sodium salt hydrate, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate hydrate
86339-1G
86339-25G
86339-5G
SIAL86339-1GEA
63.4
EUR
SIAL86339-1G
SIAL86339-25G
SIAL86339-5G
Sodium taurocholate hydrate ≥97,0% (durch TLC), Sigma-Aldrich®
Sodium taurocholate hydrate
Sodium taurocholate hydrate is used as an emulsifier in in situ preparation of digestive enzymes.
Formel:
C₂₆H₄₆NNaO₈S Molecular Weight: 555.70 g/mol |
MDL:
MFCD00150819 CAS-Nummer: 345909-26-4 |
Spezifikation Testergebnisse
Assay (TLC) | ≥97,0% |
Optical activity | [α]20/D +23±1°, c= 3% in H₂O (dry matter) |
Impurities (taurine) | ≤1% |
Impurities (cholic acid) | ≤2% |
Solubility in H₂O | 50 mg/ml clear |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...