Tetraethylammoniumtetrahydroborat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetraethylammoniumborhydrid, Tetraethylammoniumtetrahydridoborat

86600-5G
SIAL86600-5GEA 178 EUR
SIAL86600-5G
Tetraethylammoniumtetrahydroborat, Sigma-Aldrich®
Tetraethylammonium tetrahydroborate
Formel: C₈H₂₄BN
MDL: MFCD00031600
CAS-Nummer: 17083-85-1
EINECS: 241-140-4
UN: 3131
ADR: 4.3,II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR