Tetraethylammoniumnitrat, Sigma-Aldrich®
Lieferant: Merck
86645-10G
SIAL86645-10GEA
123
EUR
SIAL86645-10G
Tetraethylammoniumnitrat, Sigma-Aldrich®
Tetraethylammonium nitrate
Formel:
C₈H₂₀N₂O₃ Schmelzpunkt: > 200 °C Dichte: 1,1622 g/cm³ (25 °C) |
MDL:
MFCD00041978 CAS-Nummer: 1941-26-0 EINECS: 217-725-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...