Tetrabutylammoniumheptadecafluoroctan-1-sulfonat, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Tetrabutylammoniumperfluoroctan-1-sulfonat, Heptadecafluor-1-octansulfonsäure Tetrabutylammoniumsalz, Perfluor-1-octansulfonsäure Tetrabutylammoniumsalz

86911-5ML
SIAL86911-5MLEA 0 EUR
SIAL86911-5ML
Tetrabutylammoniumheptadecafluoroctan-1-sulfonat, Sigma-Aldrich®
Tetrabutylammoniumheptadecafluoroctan-1-sulfonat
Formel: C₂₄H₃₆F₁₇NO₃S
Molecular Weight: 741,59 g/mol
CAS-Nummer: 111873-33-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR